accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22948-94-3

IUPAC Name: 1-acetyl-1H-indole-3-carbaldehyde
Synonyms: 1-acetylindole-3-carbaldehyde
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: LCJLFGSKHBDOAY-UHFFFAOYSA-N
SMILES: CC(=O)N1C=C(C=O)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results