Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetyl-3-indolecarboxaldehyde, 98%, Thermo Scientific™

Catalog No. AC399700050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399700050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399700050 Supplier Thermo Scientific Chemicals Supplier No. 399700050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22948-94-3
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.2
MDL Number MFCD00039691
InChI Key LCJLFGSKHBDOAY-UHFFFAOYSA-N
Synonym 1-acetyl-1h-indole-3-carbaldehyde, n-acetylindole-3-carboxaldehyde, 1-acetylindole-3-carboxaldehyde, 1-acetyl-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 1-acetyl, 1-acetyl-3-formylindole, zlchem 389, pubchem7227, n-acetylindole-3-aldehyde, acmc-1crl1
PubChem CID 89915
IUPAC Name 1-acetylindole-3-carbaldehyde
SMILES CC(=O)N1C=C(C2=CC=CC=C21)C=O

Specifications

Melting Point 165°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 187.2
Percent Purity 98%
Chemical Name or Material 1-Acetyl-3-indolecarboxaldehyde
Hazard Category GHS Signal Word: Warning
EINECSNumber 245-347-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.