accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2298-13-7

IUPAC Name: hydrogen 2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]aniline chloride
Synonyms: hydrogen 4-o-tolylazo-o-toluidine chloride
Molecular Weight (g/mol): 261.75
Molecular Formula: C14H16ClN3
InChi Key: PUDCZUQFOPHIGU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC(=CC=C1N)N=NC1=CC=CC=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results