Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoazotoluene Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A058925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A058925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A058925G Supplier TCI America Supplier No. A058925G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2298-13-7
Molecular Formula C14H16ClN3
Molecular Weight (g/mol) 261.75
MDL Number MFCD00060210
InChI Key PUDCZUQFOPHIGU-UHFFFAOYSA-N
Synonym 4′C-Amino-2,3′C-dimethylazobenzene Hydrochloride
PubChem CID 16808
IUPAC Name hydrogen 2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]aniline chloride
SMILES [H+].[Cl-].CC1=CC(=CC=C1N)N=NC1=CC=CC=C1C

Specifications

Quantity 25 g
Formula Weight 261.75
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoazotoluene Hydrochloride
RTECSNumber BY6452000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.