accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3COHO

CAS RN 23001-29-8

IUPAC Name: 1',3',3'-trimethyl-1',3'-dihydrospiro[chromene-2,2'-indol]-6-ol
Synonyms: 1',3',3'-trimethylspiro[chromene-2,2'-indol]-6-ol
Molecular Weight (g/mol): 293.37
Molecular Formula: C19H19NO2
InChi Key: UHRBDHPBCHWWAG-UHFFFAOYNA-N
SMILES: CN1C2=CC=CC=C2C(C)(C)C11OC2=CC=C(O)C=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results