Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline), 99%

Catalog No. AC421920050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421920050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC421920050 Supplier Thermo Scientific Chemicals Supplier No. 421920050
  • $557.00
    $446.65 / Each of 1

    Save $110.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23001-29-8 | C19H19NO2 | 293.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23001-29-8
Molecular Formula C19H19NO2
Molecular Weight (g/mol) 293.37
MDL Number MFCD00051350
InChI Key UHRBDHPBCHWWAG-UHFFFAOYNA-N
Synonym 1',3',3'-trimethyl-6-hydroxyspiro 2h-1-benzopyran-2,2'-indoline, 1',3',3'-trimethylspiro chromene-2,2'-indolin-6-ol, spiro 2h-1-benzopyran-2,2'-indolin-6-ol, 1',3',3'-trimethyl, spiro 2h-1-benzopyran-2,2'-2h indol-6-ol, 1',3'-dihydro-1',3',3'-trimethyl, 1',3',3'-trimethylspiro chromene-2,2'-indole-6-ol, 1',3',3'-trimethylspiro chromene-2,2'-indol-6-ol, bio2g2, 1',3',3'-trimethylspiro chromene-2,2'-indoline-6-ol
PubChem CID 90866
IUPAC Name 1',3',3'-trimethylspiro[chromene-2,2'-indole]-6-ol
SMILES CN1C2=CC=CC=C2C(C)(C)C11OC2=CC=C(O)C=C2C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 149°C to 150°C
Color Brown to Purple
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 293.36
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline), 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.