accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOOONHNSOOO(Z)

CAS RN 2302-96-7

IUPAC Name: disodium 4-{2-[(1Z)-2-oxo-6-sulfonato-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}naphthalene-1-sulfonate
Synonyms: disodium 4-{2-[(1Z)-2-oxo-6-sulfonatonaphthalen-1-ylidene]hydrazin-1-yl}naphthalene-1-sulfonate
Molecular Weight (g/mol): 502.42
Molecular Formula: C20H12N2Na2O7S2
InChi Key: XYTVLRKJXNXOOI-CAPIVQQTSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(C=CC(=O)\C2=N/NC2=C3C=CC=CC3=C(C=C2)S([O-])(=O)=O)=C1
Color 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results