Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Red 13, TCI America™
SDP

Catalog No. A124310G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A124310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A124310G Supplier TCI America Supplier No. A124310G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2302-96-7
Molecular Formula C20H14N2Na2O7S2
Molecular Weight (g/mol) 504.439
MDL Number MFCD00191366
InChI Key FSSMPRHXBJPCTJ-CAPIVQQTSA-N
Synonym Fast Red E
PubChem CID 131851495
IUPAC Name (5Z)-6-oxo-5-[(4-sulfonaphthalen-1-yl)hydrazinylidene]naphthalene-2-sulfonic acid;sodium
SMILES C1=CC=C2C(=C1)C(=CC=C2S(=O)(=O)O)NN=C3C4=C(C=CC3=O)C=C(C=C4)S(=O)(=O)O.[Na].[Na]

Specifications

Color Purple/Red
Quantity 10 g
Physical Form Crystal/Powder
Formula Weight 502.42
Chemical Name or Material Acid Red 13
EINECSNumber (5)-1493
RTECSNumber QK2440000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.