accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23039-97-6

FPFCl

CAS RN 23039-97-6

IUPAC Name: chlorobis(4-fluorophenyl)phosphane
Synonyms: chlorobis(4-fluorophenyl)phosphane
Molecular Weight (g/mol): 256.62
Molecular Formula: C12H8ClF2P
InChi Key: PYFWYPBHSZATST-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)P(Cl)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results