Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobis(4-fluorophenyl)phosphine, 98%

Catalog No. AAH26953MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26953MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH26953MD Supplier Thermo Scientific Chemicals Supplier No. H26953MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 23039-97-6 | C12H8ClF2P | 256.617 g/mol

In chemical research. Used as an interphasic transfer catalyst, a polymerization catalyst, and a trans-halogenation catalyst. As a precursor to fire-retardant materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In chemical research. Used as an interphasic transfer catalyst, a polymerization catalyst, and a trans-halogenation catalyst. As a precursor to fire-retardant materials.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23039-97-6
Molecular Formula C12H8ClF2P
Molecular Weight (g/mol) 256.617
MDL Number MFCD04972302
InChI Key PYFWYPBHSZATST-UHFFFAOYSA-N
Synonym bis 4-fluorophenyl chlorophosphine, chlorobis 4-fluorophenyl phosphine, chloro-bis 4-fluorophenyl phosphane, chlorobis 4-fluorophenyl phosphane, acmc-20alnm, bis-4-fluorophenyl chlorophosphine, phosphinous chloride,p,p-bis 4-fluorophenyl
PubChem CID 4379931
IUPAC Name chloro-bis(4-fluorophenyl)phosphane
SMILES C1=CC(=CC=C1F)P(C2=CC=C(C=C2)F)Cl

Specifications

Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 256.62
Percent Purity 98%
Chemical Name or Material Chlorobis(4-fluorophenyl)phosphine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.