accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NONHOOOHO

CAS RN 2304-96-3

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-carbamoylpropanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3-carbamoylpropanoic acid
Molecular Weight (g/mol): 266.25
Molecular Formula: C12H14N2O5
InChi Key: FUCKRCGERFLLHP-UHFFFAOYNA-N
SMILES: NC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)