Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Carbobenzoxy-L-asparagine 99.0+%, TCI America™
SDP

Catalog No. C05731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05731G 1 g
C057310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C05731G Supplier TCI America Supplier No. C05731G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2304-96-3
Molecular Formula C12H14N2O5
Molecular Weight (g/mol) 266.253
MDL Number MFCD00008035
InChI Key FUCKRCGERFLLHP-VIFPVBQESA-N
Synonym z-asn-oh, n-cbz-l-asparagine, cbz-asn-oh, cbz asparagine, n-benzyloxycarbonyl-l-asparagine, n-carbobenzoxy-l-asparagine, z-l-asparagine, carbobenzyloxy-l-asparagine, carbobenzoxy-l-asparagin
PubChem CID 75314
IUPAC Name (2S)-4-amino-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC(=O)N)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 266.25
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-Carbobenzoxy-L-asparagine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.