accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HNNOONNHOOOHO(E/Z)

CAS RN 2304-98-5

IUPAC Name: 5-{[amino(nitroamino)methylidene]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonyms: 5-{[amino(nitroamino)methylidene]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 353.34
Molecular Formula: C14H19N5O6
InChi Key: BZPCSFNCKORLQG-UHFFFAOYNA-N
SMILES: NC(N[N+]([O-])=O)=NCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)