accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23056-36-2

NNOOCl

CAS RN 23056-36-2

IUPAC Name: 2-chloro-4-nitropyridine
Synonyms: 2-chloro-4-nitropyridine
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: LIEPVGBDUYKPLC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=NC=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)