accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 23074-42-2

IUPAC Name: adamantane-1-carbonitrile
Synonyms: adamantane-1-carbonitrile
Molecular Weight (g/mol): 161.25
Molecular Formula: C11H15N
InChi Key: FQFZASRJFRAEIH-UHFFFAOYSA-N
SMILES: N#CC12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)