Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adamantane-1-carbonitrile, 97%

Catalog No. AAA1435206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1435206 5 g
AAA1435203 1 g
AAA1435214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1435206 Supplier Thermo Scientific Chemicals Supplier No. A1435206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 23074-42-2 | C11H15N | 161.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23074-42-2
Molecular Formula C11H15N
Molecular Weight (g/mol) 161.248
MDL Number MFCD00074731
InChI Key FQFZASRJFRAEIH-UHFFFAOYSA-N
Synonym 1-adamantanecarbonitrile, 1-cyanoadamantane, 1-adamantyl cyanide, tricyclo 3.3.1.13,7 decane-1-carbonitrile, tricyclo 3.3.1.1∼3,7∼ decane-1-carbonitrile, adamantanecarbonitrile, 3r,5s,7s-adamantane-1-carbonitrile, 1-adamantylnitrile, pubchem8757
PubChem CID 90878
IUPAC Name adamantane-1-carbonitrile
SMILES C1C2CC3CC1CC(C2)(C3)C#N

Specifications

Melting Point 193°C to 196°C
Quantity 5 g
UN Number UN3439
Formula Weight 161.25
Percent Purity 97%
Chemical Name or Material Adamantane-1-carbonitrile
Hazard Category H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 245-413-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.