accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NHN

CAS RN 23099-21-0

IUPAC Name: 1-(piperidin-3-yl)methanamine
Synonyms: 1-(piperidin-3-yl)methanamine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: IPOVLZSJBYKHHU-UHFFFAOYNA-N
SMILES: NCC1CCCNC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results