Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Aminomethyl)piperidine 98.0+%, TCI America™
SDP

Catalog No. A25185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25185G Supplier TCI America Supplier No. A25185G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23099-21-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD03427344
InChI Key IPOVLZSJBYKHHU-UHFFFAOYSA-N
PubChem CID 90880
IUPAC Name piperidin-3-ylmethanamine
SMILES C1CC(CNC1)CN

Specifications

Color Colorless
Boiling Point 89°C
Quantity 5 g
UN Number 2735
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Aminomethyl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.