accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNN

CAS RN 231-23-2

IUPAC Name: pyrazino[2,3-f]quinoxaline
Synonyms: pyrazino[2,3-f]quinoxaline
Molecular Weight (g/mol): 182.19
Molecular Formula: C10H6N4
InChi Key: CVSGFMWKZVZOJD-UHFFFAOYSA-N
SMILES: C1=CN=C2C(C=CC3=NC=CN=C23)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results