Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazino[2,3-f]quinoxaline, 99+%, Thermo Scientific™

Catalog No. AC252862500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC252862500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC252862500 Supplier Thermo Scientific Chemicals Supplier No. 252862500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 231-23-2
Molecular Formula C10H6N4
Molecular Weight (g/mol) 182.186
InChI Key CVSGFMWKZVZOJD-UHFFFAOYSA-N
Synonym pyrazino 2,3-f quinoxaline, 1,4,5,8-tetraazaphenanthrene, pyrazino 2,3-f quinoxaline #, 1,4,5,8-tetraaza-phenanthrene, 1,4,5,8-tetra-aza-phenathrene, 4,7-diaza-1,10-phenanthroline
PubChem CID 174669
IUPAC Name pyrazino[2,3-f]quinoxaline
SMILES C1=CC2=NC=CN=C2C3=NC=CN=C31

Specifications

Melting Point 249°C to 252°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 250 mg
Formula Weight 182.18
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material Pyrazino[2, 3-f]quinoxaline, 99+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.