accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPSOCH3SNOOCl

CAS RN 2310-17-0

IUPAC Name: O,O-diethyl {[(6-chloro-2-oxo-2,3-dihydro-1,3-benzoxazol-3-yl)methyl]sulfanyl}phosphonothioate
Synonyms: phosalone phosalone (substance) chipman 11974 fozalone benzphos benzophosphate caswell no. 660A zolone DT zolone PM phoazlone
Molecular Weight (g/mol): 367.8
Molecular Formula: C12H15ClNO4PS2
InChi Key: IOUNQDKNJZEDEP-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCN1C(=O)OC2=CC(Cl)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results