accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(S)(R)(R)O(S)(R)CH3OH3CCH3OOOHO(E)(E)(E)(E)

CAS RN 23110-15-8

IUPAC Name: (2E,4E,6E,8E)-10-{[(3R,4S,5S,6R)-5-methoxy-4-[(2R)-2-methyl-3-(3-methylbut-2-en-1-yl)oxiran-2-yl]-1-oxaspiro[2.5]octan-6-yl]oxy}-10-oxodeca-2,4,6,8-tetraenoic acid
Synonyms: fumidil fumagilline fumagillina fumidil B fumagillin dch fumagillin (substance) amebacillin fugillin fumagillinum fugilin
Molecular Weight (g/mol): 458.55
Molecular Formula: C26H34O7
InChi Key: NGGMYCMLYOUNGM-IJIYPQOHSA-N
SMILES: CO[C@@H]1[C@@H](CC[C@]2(CO2)[C@H]1[C@@]1(C)OC1CC=C(C)C)OC(=O)\C=C\C=C\C=C\C=C\C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results