Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Fumagillin, Aspergillus fumigatus

Catalog No. 344845500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-484-5500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-484-5500UG Supplier MilliporeSigma™ Supplier No. 344845500UG
Add to Cart
Add to Cart

Antiprotozoal agent and angiogenesis inhibitor.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 90% by HPLC
Target Proteins Methionine amino-peptidase 2
CAS 23110-15-8
Molecular Formula C26H34O7
Formula Weight 458.6g/mol
Solubility Aqueous Alkali or DMSO
Content And Storage Protect from light
Form Solid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.