accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2312-14-3

IUPAC Name: 4-(dodecyloxy)-N-{[4-(dodecyloxy)phenyl]imino}benzen-1-imine oxide
Synonyms: 4-(dodecyloxy)-N-{[4-(dodecyloxy)phenyl]imino}benzenimine oxide
Molecular Weight (g/mol): 566.87
Molecular Formula: C36H58N2O3
InChi Key: RFLXFGRPXYHTKL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC1=CC=C(C=C1)N=[N+]([O-])C1=CC=C(OCCCCCCCCCCCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results