Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Didodecyloxyazoxybenzene 98.0+%, TCI America™
SDP

Catalog No. D05535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D05535G Supplier TCI America Supplier No. D05535G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2312-14-3
Molecular Formula C36H58N2O3
Molecular Weight (g/mol) 566.87
MDL Number MFCD00059283
InChI Key RFLXFGRPXYHTKL-UHFFFAOYSA-N
PubChem CID 11489953
IUPAC Name 4-(dodecyloxy)-N-{[4-(dodecyloxy)phenyl]imino}benzen-1-imine oxide
SMILES CCCCCCCCCCCCOC1=CC=C(C=C1)N=[N+]([O-])C1=CC=C(OCCCCCCCCCCCC)C=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 566.87
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Didodecyloxyazoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.