accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2312-23-4

IUPAC Name: hydrogen (3-chlorophenyl)hydrazine chloride
Synonyms: hydrogen (3-chlorophenyl)hydrazine chloride
Molecular Weight (g/mol): 179.04
Molecular Formula: C6H8Cl2N2
InChi Key: CRRIAWUJYMLJOE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (5)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)