accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3ONNNH2H2N(E/Z)

CAS RN 2313-87-3

IUPAC Name: hydrogen 4-[2-(4-ethoxyphenyl)diazen-1-yl]benzene-1,3-diamine chloride
Synonyms: hydrogen 4-[2-(4-ethoxyphenyl)diazen-1-yl]benzene-1,3-diamine chloride
Molecular Weight (g/mol): 292.77
Molecular Formula: C14H17ClN4O
InChi Key: IWHXNINOLLNFGP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results