Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxychrysoidine Hydrochloride 95.0+%, TCI America™
SDP

Catalog No. E00545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00545G 5 g
E005425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E00545G Supplier TCI America Supplier No. E00545G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2313-87-3
Molecular Formula C14H17ClN4O
Molecular Weight (g/mol) 292.77
MDL Number MFCD00042023
InChI Key IWHXNINOLLNFGP-UHFFFAOYSA-N
Synonym 4-(4-Ethoxyphenylazo)-1,3-phenylenediamine Monohydrochloride, Ethoxy Red
PubChem CID 16836
IUPAC Name hydrogen 4-[2-(4-ethoxyphenyl)diazen-1-yl]benzene-1,3-diamine chloride
SMILES [H+].[Cl-].CCOC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1N

Specifications

Color Black-Yellow
Quantity 5 g
Formula Weight 292.77
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxychrysoidine Hydrochloride
RTECSNumber ST2510000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.