accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23145-88-2

IUPAC Name: 1-[(4-chlorophenyl)methyl]piperazine
Synonyms: 1-[(4-chlorophenyl)methyl]piperazine
Molecular Weight (g/mol): 210.71
Molecular Formula: C11H15ClN2
InChi Key: GSJXJZOWHSTWOX-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CN2CCNCC2)C=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)