accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23222-97-1

IUPAC Name: [4-(hydroxymethyl)-2-nitrophenyl]methanol
Synonyms: [4-(hydroxymethyl)-2-nitrophenyl]methanol
Molecular Weight (g/mol): 183.16
Molecular Formula: C8H9NO4
InChi Key: KWVHOBYJXDKIPL-UHFFFAOYSA-N
SMILES: OCC1=CC=C(CO)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results