accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOHOOH3CHOHO

CAS RN 23235-61-2

IUPAC Name: 2-ethyl-2-{[2-ethyl-3-hydroxy-2-(hydroxymethyl)propoxy]methyl}propane-1,3-diol
Synonyms: DI(trimethylolpropane)
Molecular Weight (g/mol): 250.34
Molecular Formula: C12H26O5
InChi Key: WMYINDVYGQKYMI-UHFFFAOYSA-N
SMILES: CCC(CO)(CO)COCC(CC)(CO)CO
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)