accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONSNNHN

CAS RN 23249-95-8

IUPAC Name: 4-(5-sulfanylidene-2,5-dihydro-1H-1,2,3,4-tetrazol-1-yl)benzoic acid
Synonyms: 4-(5-sulfanylidene-2H-1,2,3,4-tetrazol-1-yl)benzoic acid
Molecular Weight (g/mol): 222.22
Molecular Formula: C8H6N4O2S
InChi Key: GDVFHEXRJFFDDB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)N1NN=NC1=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results