Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Carboxyphenyl)-5-mercapto-1H-tetrazole 95.0+%, TCI America™
SDP

Catalog No. C20115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20115G Supplier TCI America Supplier No. C20115G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23249-95-8
Molecular Formula C8H6N4O2S
Molecular Weight (g/mol) 222.22
MDL Number MFCD01074835
InChI Key GDVFHEXRJFFDDB-UHFFFAOYSA-N
Synonym 1-(4-Carboxyphenyl)-1H-tetrazole-5-thiol, 4-(5-Mercapto-1H-tetrazol-1-yl)benzoic Acid
PubChem CID 13651022
IUPAC Name 4-(5-sulfanylidene-2,5-dihydro-1H-1,2,3,4-tetrazol-1-yl)benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)N1NN=NC1=S

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1325
Formula Weight 222.22
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Carboxyphenyl)-5-mercapto-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.