accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23256-33-9

IUPAC Name: dihydrogen [3-(carbamimidoylsulfanyl)propyl]dimethylamine dichloride
Synonyms: dimaprit
Molecular Weight (g/mol): 234.18
Molecular Formula: C6H17Cl2N3S
InChi Key: DFWCPLGXFMSUCW-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CN(C)CCCSC(N)=N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results