Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimaprit dihydrochloride, R&D Systems™
SDP

Catalog No. 0506100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-061-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 05-061-00 Supplier R&D Systems Supplier No. 0506/100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Standard H2 selective agonist

Selective H2 agonist with central effects upon systemic administration. Also inhibits nNOS (IC50 = 49 μM).

Chemical Identifiers

CAS 23256-33-9
Molecular Formula C6H17Cl2N3S
Molecular Weight (g/mol) 234.183
InChI Key DFWCPLGXFMSUCW-UHFFFAOYSA-N
Synonym dimaprit dihydrochloride, diamaprit-2hcl, dimaprit hydrochloride, dimaprit-2hcl, usaf pd-40, 2-3'-dimethylaminopropylthio pseudourea dihydrochloride, 2-3-dimethylaminopropyl-2-thiopseudourea dihydrochloride, 3-dimethylamino propyl carbamimidothioate dihydrochloride, 3-carbamimidoylsulfanyl propyl dimethylamine dihydrochloride, pseudourea, 2-3-dimethylamino propyl-2-thio-, dihydrochloride
PubChem CID 90045
IUPAC Name 3-(dimethylamino)propyl carbamimidothioate;dihydrochloride
SMILES CN(C)CCCSC(=N)N.Cl.Cl

Specifications

Quantity 100 mg
Formula Weight 234.19
Percent Purity >99%
Product Line Tocris
Chemical Name or Material Dimaprit dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.