accessibility menu, dialog, popup

UserName

Viewing profile as user:
PSBr

CAS RN 23259-98-5

IUPAC Name: triphenyl[(thiophen-2-yl)methyl]phosphanium bromide
Synonyms: triphenyl(thiophen-2-ylmethyl)phosphanium bromide
Molecular Weight (g/mol): 439.35
Molecular Formula: C23H20BrPS
InChi Key: PJQVALHFZIYJMT-UHFFFAOYSA-M
SMILES: [Br-].C(C1=CC=CS1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results