Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenyl(2-thienylmethyl)phosphonium Bromide 98.0+%, TCI America™
SDP

Catalog No. T31591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T31591G Supplier TCI America Supplier No. T31591G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23259-98-5
Molecular Formula C23H20BrPS
Molecular Weight (g/mol) 439.35
MDL Number MFCD03931091
InChI Key PJQVALHFZIYJMT-UHFFFAOYSA-M
PubChem CID 10741697
IUPAC Name triphenyl[(thiophen-2-yl)methyl]phosphanium bromide
SMILES [Br-].C(C1=CC=CS1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 275°C
Quantity 1 g
Formula Weight 439.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenyl(2-thienylmethyl)phosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.