accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23282-55-5

IUPAC Name: sodium (4-aminophenyl)[(6-chloropyridazin-3-yl)imino]-λ⁶-sulfanoylolate
Synonyms: sodium (4-aminophenyl)[(6-chloropyridazin-3-yl)imino]-λ⁶-sulfanoylolate
Molecular Weight (g/mol): 306.7
Molecular Formula: C10H8ClN4NaO2S
InChi Key: QUJXXPVBXOUFDH-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC=C(C=C1)S([O-])(=O)=NC1=CC=C(Cl)N=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results