Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfachloropyridazine sodium, 97%, Thermo Scientific™

Catalog No. AC458060050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458060050 5 g
AC458060010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458060050 Supplier Thermo Scientific Chemicals Supplier No. 458060050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23282-55-5
Molecular Formula C10H8ClN4NaO2S
Molecular Weight (g/mol) 306.7
InChI Key GHQJXOBLWKFNAD-UHFFFAOYSA-N
PubChem CID 87077948
IUPAC Name 4-amino-N-(6-chloropyridazin-3-yl)benzenesulfonamide;sodium
SMILES C1=CC(=CC=C1N)S(=O)(=O)NC2=NN=C(C=C2)Cl.[Na]

Specifications

Melting Point 262.0°C to 270.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 306.7
Percent Purity 97%
Chemical Name or Material Sulfachloropyridazine sodium
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 245-553-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.