accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23299-85-6

IUPAC Name: 1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one copper
Synonyms: 1-[2-(pyridin-2-yl)hydrazin-1-ylidene]naphthalen-2-one copper
Molecular Weight (g/mol): 312.82
Molecular Formula: C15H11CuN3O
InChi Key: MBXMXGGKDLXUGT-UHFFFAOYSA-N
SMILES: [Cu].O=C1C=CC2=CC=CC=C2C1=NNC1=CC=CC=N1
Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results