Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper PAN, TCI America™
SDP

Catalog No. C03851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C03851G Supplier TCI America Supplier No. C03851G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23299-85-6
Molecular Formula C15H11CuN3O
Molecular Weight (g/mol) 312.82
MDL Number MFCD00067176
InChI Key MBXMXGGKDLXUGT-UHFFFAOYSA-N
Synonym Bis[1-(2-pyridylazo)-2-naphtholato]copper(II)
PubChem CID 130424121
IUPAC Name 1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one copper
SMILES [Cu].O=C1C=CC2=CC=CC=C2C1=NNC1=CC=CC=N1

Specifications

Quantity 1 g
Form Crystalline Powder
Chemical Name or Material Copper PAN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.