accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNOOCH3

CAS RN 23315-55-1

IUPAC Name: 2-oxo-1,2-bis(4-propoxyphenyl)hydrazin-2-ium-1-ide
Synonyms: 2-oxo-1,2-bis(4-propoxyphenyl)hydrazin-2-ium-1-ide
Molecular Weight (g/mol): 314.39
Molecular Formula: C18H22N2O3
InChi Key: YRZFKJUPVCUDMX-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C([N-][N+](=O)C2=CC=C(OCCC)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results