Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dipropoxyazoxybenzene 98.0+%, TCI America™
SDP

Catalog No. D10945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D10945G Supplier TCI America Supplier No. D10945G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23315-55-1
Molecular Formula C18H22N2O3
Molecular Weight (g/mol) 314.385
MDL Number MFCD00059870
InChI Key YRZFKJUPVCUDMX-UHFFFAOYSA-N
PubChem CID 90063
IUPAC Name oxido-(4-propoxyphenyl)-(4-propoxyphenyl)iminoazanium
SMILES CCCOC1=CC=C(C=C1)N=[N+](C2=CC=C(C=C2)OCCC)[O-]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 314.39
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dipropoxyazoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.