accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH(S)OH(R)OH(R)OH(R)OHOH

CAS RN 23323-37-7

IUPAC Name: (2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol
Synonyms: (2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol
Molecular Weight (g/mol): 293.4
Molecular Formula: C14H31NO5
InChi Key: ZRRNJJURLBXWLL-REWJHTLYSA-N
SMILES: CCCCCCCCNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)