accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOON

CAS RN 23353-14-2

IUPAC Name: 2-[2-(4-methoxyphenyl)-1,3-thiazol-4-yl]acetate
Synonyms: 2-[2-(4-methoxyphenyl)-1,3-thiazol-4-yl]acetate
Molecular Weight (g/mol): 248.28
Molecular Formula: C12H10NO3S
InChi Key: HWRQFRXTBPYUEK-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1)C1=NC(CC([O-])=O)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results