accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClSClOO

CAS RN 23378-88-3

IUPAC Name: 3,5-dichloro-2-hydroxybenzene-1-sulfonyl chloride
Synonyms: 3,5-dichloro-2-hydroxybenzenesulfonyl chloride
Molecular Weight (g/mol): 261.5
Molecular Formula: C6H3Cl3O3S
InChi Key: KXFQRJNVGBIDHA-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(Cl)C=C1S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)