accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OMgOOO

CAS RN 23389-33-5

IUPAC Name: magnesium(2+) hydrate carbonate
Synonyms: magnesium(2+) hydrate carbonate
Molecular Weight (g/mol): 102.33
Molecular Formula: CH2MgO4
InChi Key: OUHCLAKJJGMPSW-UHFFFAOYSA-L
SMILES: O.[Mg++].[O-]C([O-])=O
Grade 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)