Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Magnesium Carbonate, USP (Heavy)
SDP

Catalog No. 14185592 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-592 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-592 Supplier Medisca Supplier No. 140508
Add to Cart
Add to Cart

Magnesium Carbonate, USP (Heavy) varies between light, white, friable masses or bulky, white powder.

Magnesium Carbonate, USP (Heavy) varies between light, white, friable masses or bulky, white powder. It is practically insoluble in water to which, however, it imparts a slightly alkaline reaction.

Alternative name(s): Magnesium carbonate Hydroxide Magnesite

Specifications

Chemical Name or Material Magnesium Carbonate, USP (Heavy)
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 23389-33-5
Type Drug Standard
Molecular Weight (g/mol) 84.31 (anhydre); 485.65 (base); 102.33 (monohydrate)
Quantity 500 g
Percent Purity 40 to 43.5 % (magnesium oxide)
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.