accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 23391-99-3

IUPAC Name: [1,1'-bi(cyclohexane)]-4,4'-dione
Synonyms: [1,1'-bi(cyclohexane)]-4,4'-dione
Molecular Weight (g/mol): 194.27
Molecular Formula: C12H18O2
InChi Key: OQIDYCVWAFZRTR-UHFFFAOYSA-N
SMILES: O=C1CCC(CC1)C1CCC(=O)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results