Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bicyclohexanone 98.0+%, TCI America™
SDP

Catalog No. B2405500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2405500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2405500G Supplier TCI America Supplier No. B2405500G

Chemical Identifiers

CAS 23391-99-3
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD00210694
InChI Key OQIDYCVWAFZRTR-UHFFFAOYSA-N
PubChem CID 264825
IUPAC Name [1,1'-bi(cyclohexane)]-4,4'-dione
SMILES O=C1CCC(CC1)C1CCC(=O)CC1

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 500 g
Synonym 4,4′-Dioxobicyclohexyl
Formula Weight 194.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bicyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.